Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(-)-1,1'-Binaphthyl-2,2'-diyl Hydrogen Phosphate 98.0+%, TCI America™
SDP

Catalog No. B11431G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B11431G 1 g
B1143100MG 100 mg
B11435G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B11431G Supplier TCI America Supplier No. B11431G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 39648-67-4
Molecular Formula C20H12O4P
Molecular Weight (g/mol) 347.29
MDL Number MFCD00010045
InChI Key JEHUZVBIUCAMRZ-UHFFFAOYSA-M
Synonym 1,1'-binaphthyl-2,2'-diyl hydrogenphosphate, s-+-1,1'-binaphthyl-2,2'-diyl hydrogenphosphate, r---1,1'-binaphthyl-2,2'-diyl hydrogenphosphate, 1,1'-binaphthyl-2,2'-diyl hydrogen phosphate, r---bnp acid, 11bs-4-hydroxydinaphtho 2,1-d:1',2'-f 1,3,2 dioxaphosphepine 4-oxide, r---1,1'-binaphthyl-2,2'-diyl hydrogen phosphate, r-4-hydroxydinaphtho 2,1-d:1',2'-f 1,3,2 dioxaphosphepine 4-oxide, s-+-bnp acid, 4-hydroxydinaphtho 2,1-d:1',2'-f 1,3,2 dioxaphosphepine 4-oxide
PubChem CID 99589
IUPAC Name 13-oxo-12,14-dioxa-13λ⁵-phosphapentacyclo[13.8.0.0²,¹¹.0³,⁸.0¹⁸,²³]tricosa-1(23),2,4,6,8,10,15,17,19,21-decaen-13-olate
SMILES [O-]P1(=O)OC2=CC=C3C=CC=CC3=C2C2=C3C=CC=CC3=CC=C2O1

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 348.29
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (R)-(-)-1,1′-Binaphthyl-2,2′-diyl Hydrogen Phosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.