Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-1,2-Butanediol 98.0+%, TCI America™
SDP

Catalog No. B41181G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B41181G 1 g
B41185G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B41181G Supplier TCI America Supplier No. B41181G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 40348-66-1
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.12
MDL Number MFCD09953765
InChI Key BMRWNKZVCUKKSR-UHFFFAOYNA-N
Synonym (R)-1,2-Butylene Glycol, (R)-1,2-Dihydroxybutane
PubChem CID 641012
ChEBI CHEBI:52685
IUPAC Name butane-1,2-diol
SMILES CCC(O)CO

Specifications

Color Yellow
Boiling Point 60°C
Quantity 1 g
Formula Weight 90.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (R)-1,2-Butanediol
EINECSNumber (2)-0235
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.