Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(+)-1-(4-Methoxyphenyl)ethylamine 98.0+%, TCI America™
SDP

Catalog No. M151125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M151125G 25 g
M15115G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M151125G Supplier TCI America Supplier No. M151125G

Chemical Identifiers

CAS 22038-86-4
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.209
MDL Number MFCD00671659
InChI Key JTDGKQNNPKXKII-SSDOTTSWSA-N
Synonym r-+-1-4-methoxyphenyl ethylamine, r-1-4-methoxyphenyl ethylamine, r-1-4-methoxyphenyl ethanamine, 1r-1-4-methoxyphenyl ethanamine, r-+-4-methoxy-alpha-methylbenzylamine, s-4-methoxy-alpha-methylbenzylamine, r +-1-4-methylphenyl ethylamine, pubchem9920, ksc496q8b, rm+-1-4-methoxyphenyl ethylamine
PubChem CID 641449
IUPAC Name (1R)-1-(4-methoxyphenyl)ethanamine
SMILES CC(C1=CC=C(C=C1)OC)N

Specifications

Color Yellow
pH 11 (10g/L H2O soln. 20°C)
Quantity 25 g
UN Number 2735
Formula Weight 151.21
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material (R)-(+)-1-(4-Methoxyphenyl)ethylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.