Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(-)-1-Cyclohexylethylamine 98.0+%, TCI America™
SDP

Catalog No. C154125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C154125G 25 g
C15415G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C154125G Supplier TCI America Supplier No. C154125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5913-13-3
Molecular Formula C8H17N
Molecular Weight (g/mol) 127.23
MDL Number MFCD00043338
InChI Key XBWOPGDJMAJJDG-UHFFFAOYNA-N
Synonym r---1-cyclohexylethylamine, r-1-cyclohexylethylamine, 1r-1-cyclohexylethanamine, r-1-cyclohexylethanamine, 1r-1-cyclohexylethylamine, chembl19290, 1r-1-cyclohexylethan-1-amine, 1-cyclohexyl-ethylamine, 1-cyclohexylethanamine #, r---cyclohexylethylamine
PubChem CID 10997046
IUPAC Name 1-cyclohexylethan-1-amine
SMILES CC(N)C1CCCCC1

Specifications

Color Yellow
Boiling Point 60°C
Quantity 25 g
UN Number 2734
Formula Weight 127.23
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material (R)-(-)-1-Cyclohexylethylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.