Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(+)-1-Fmoc-4-oxopiperidine-2-carboxylic acid, 97%, Thermo Scientific™

Catalog No. AAH6632803
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6632803 1 g
AAH66328MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6632803 Supplier Thermo Scientific Chemicals Supplier No. H6632803

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1375078-38-8
Molecular Formula C21H19NO5
Molecular Weight (g/mol) 365.385
MDL Number MFCD22208545
InChI Key YGAHCGORXONSSM-LJQANCHMSA-N
Synonym r-1-fmoc-4-oxopiperidine-2-carboxylic acid, 2r-1-9h-fluoren-9-ylmethoxy carbonyl-4-oxopiperidine-2-carboxylic acid
PubChem CID 92174274
IUPAC Name (2R)-1-(9H-fluoren-9-ylmethoxycarbonyl)-4-oxopiperidine-2-carboxylic acid
SMILES C1CN(C(CC1=O)C(=O)O)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24

Specifications

Color White to Yellow
Quantity 1 g
Sensitivity Moisture sensitive
Formula Weight 365.38
Percent Purity 97%
Physical Form Powder
Chemical Name or Material (R)-(+)-1-Fmoc-4-oxopiperidine-2-carboxylic acid
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold; Store under Nitrogen
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.