Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(+)-1-Phenylethylamine, 99+%

Catalog No. p-2689245
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC371460500 50 g Glass bottle
AC371462500 250 g Glass bottle
AC371460010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC371460500 Supplier Thermo Scientific Chemicals Supplier No. 371460500
Only null left
Add to Cart

CAS: 3886-69-9 | C8H11N | 121.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chiral derivatization reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3886-69-9
MDL Number MFCD00064405
InChI Key RQEUFEKYXDPUSK-SSDOTTSWSA-N
Synonym r-1-phenylethanamine, r-+-1-phenylethylamine, 1r-1-phenylethanamine, r-1-phenylethylamine, d-alpha-methylbenzylamine, 1r-1-phenylethan-1-amine, r-+-alpha-methylbenzylamine, r-alpha-methylbenzenemethanamine, unii-v022zk8gz5, r-alfa-methylbenzylamine
PubChem CID 643189
ChEBI CHEBI:35322
IUPAC Name (1R)-1-phenylethanamine
SMILES CC(C1=CC=CC=C1)N

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -10.0°C
Color Colorless
Density 0.9500g/mL
Boiling Point 184.0°C to 186.0°C
Flash Point 75°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Molecular Formula C8H11N
Refractive Index 1.5250 to 1.5270
Linear Formula C6H5CH(CH3)NH2
Quantity 50 g
Beilstein 12,II,586
Fieser 09,363; 03,199; 14,257; 13,185; 12,319; 11,411; 07,393; 05,441; 02,271; 01,838; 06,457
Merck Index 13,6054
Specific Gravity 0.95
Specific Rotation Condition + 40.00 (20.00°C neat)
Specific Rotation + 40.00
Solubility Information Solubility in water: 40g/L water (20°C). Other solubilities: soluble in most organic solvents,miscible with alcohols and ethers
Molecular Weight (g/mol) 121.18
Formula Weight 121.18
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material (R)-(+)-1-Phenylethylamine, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 223-423-4

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.