Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(+)-2-Chloropropionic Acid 98.0+%, TCI America™
SDP

Catalog No. C165525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C165525G 25 g
C16551G 1 g
C16555G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C165525G Supplier TCI America Supplier No. C165525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7474-05-7
Molecular Formula C3H4ClO2
Molecular Weight (g/mol) 107.51
MDL Number MFCD00064204
InChI Key GAWAYYRQGQZKCR-UWTATZPHSA-M
Synonym r-+-2-chloropropionic acid, r-2-chloropropanoic acid, r-2-chloropropionic acid, r-2-chloro-propionic acid, r +-2-chloropropionic acid, unii-f60st8319r, 2-chloropropionic acid, +, propanoic acid, 2-chloro-, 2r, d-2-chloropropionic acid, +-2-chloropropanoic acid
PubChem CID 2724540
IUPAC Name (2R)-2-chloropropanoic acid
SMILES CC(C(=O)O)Cl

Specifications

Color Colorless
Quantity 25 g
UN Number 2511
Formula Weight 108.52
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material (R)-(+)-2-Chloropropionic Acid
EINECSNumber (2)-1157
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.