Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(-)-2-Methylpiperazine, 98%, Acros Organics™

Click to view available options
Quantity:
1g
5g
25g
75336-86-6
C5H12N2
100.16
JOMNTHCQHJPVAZ-RXMQYKEDSA-N
r---2-methylpiperazine,r-2-methylpiperazine,2r-2-methylpiperazine,r-mp,piperazine, 2-methyl-, 2r,r-2-methyl-piperazine,2-methylpiperazine, r,unii-p4xu643r8w
7330434
(2R)-2-methylpiperazine
CC1CNCCN1
75336-86-6
C5H12N2
100.16
JOMNTHCQHJPVAZ-RXMQYKEDSA-N
r---2-methylpiperazine,r-2-methylpiperazine,2r-2-methylpiperazine,r-mp,piperazine, 2-methyl-, 2r,r-2-methyl-piperazine,2-methylpiperazine, r,unii-p4xu643r8w
7330434
(2R)-2-methylpiperazine
CC1CNCCN1
Boiling Point 155.0°C to 156.0°C
Specific Rotation Condition − 18.00 (20.00°C c=1,toluene)
Melting Point 86.0°C to 91.0°C
Infrared Spectrum Authentic
Color White to Yellow
Quantity 25g
Flash Point 65°C
Packaging Glass bottle
Specific Rotation − 18.00
Assay Percent Range 97.5% min. (GC)
Solubility Information Solubility in water: soluble.
Formula Weight 100.16
Physical Form Crystalline Powder, Crystals and/or Chunks
Percent Purity 98%
Chemical Name or Material (R)-(-)-2-Methylpiperazine
Show More Show Less

GHS Signal Word: Danger