Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(-)-2-Methylpyrrolidine, 95%

Catalog No. AAH5788503
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5788503 1 g
AAH57885MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5788503 Supplier Thermo Scientific Chemicals Supplier No. H5788503
Add to Cart

CAS: 41720-98-3 | C5H11N | 85.15 g/mol

It is used as a building block used in a multikilogram synthesis of a chiral naphthalenoid histamine receptor antagonist.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a building block used in a multikilogram synthesis of a chiral naphthalenoid histamine receptor antagonist.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Keep container in a cool refrigerated, well-ventilated area. Moisture sensitive
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41720-98-3
Molecular Formula C5H11N
Molecular Weight (g/mol) 85.15
MDL Number MFCD07783026
InChI Key RGHPCLZJAFCTIK-RXMQYKEDSA-N
Synonym r-2-methylpyrrolidine, r-2-methyl-pyrrolidine, 2r-2-methylpyrrolidine, r---2-methylpyrrolidine, r-2-methyl pyrrolidine, 2-r-methylpyrrolidine, 2r---2-methyltetrahydropyrrole, 2r---2-methylpyrrolidine, pyrrolidine, 2-methyl-, 2r, 2-methyl-pyrrolidine
PubChem CID 641544
ChEBI CHEBI:78858
IUPAC Name (2R)-2-methylpyrrolidine
SMILES CC1CCCN1

Specifications

Density 0.84
Flash Point 7°C (45°F)
Refractive Index 1.43
Quantity 1 g
UN Number UN2733
Solubility Information Insoluble in water.
Optical Rotation −19° (c=5 in Methanol)
Formula Weight 85.15
Percent Purity 95%
Chemical Name or Material (R)-(-)-2-Methylpyrrolidine
Hazard Category H225-H314-H335
Hazard Statement GHS H Statement
H224-H314-H318
Extremely flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.