Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(-)-2-Pentanol, 97%, Thermo Scientific™

Catalog No. AAJ63298MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63298MD 250 mg
AAJ6329803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ63298MD Supplier Thermo Scientific Chemicals Supplier No. J63298MD

missing translation for 'validMainframeMsg'

For synthesis of optically active products

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31087-44-2
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00065953
InChI Key JYVLIDXNZAXMDK-RXMQYKEDSA-N
Synonym r---2-pentanol, 2r-pentan-2-ol, r-2-pentanol, 2r-2-pentanol, 2-pentanol #, r-pentan-2-ol, pubchem6721, 2-pentanol, 2r
PubChem CID 7014876
IUPAC Name (2R)-pentan-2-ol
SMILES CCCC(C)O

Specifications

Color Colorless
Density 0.81
Boiling Point 119°C to 120°C
Flash Point 34°C (93°F)
Refractive Index 1.406
Quantity 250 mg
UN Number UN1105
Beilstein 4652311
Optical Rotation −14° (Neat)
Formula Weight 88.15
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material (R)-(-)-2-Pentanol
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H226-H332-H335
Flammable liquid and vapor.
Harmful if inhaled.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P210-P261-P280-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: PENTANOLS
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.