Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-3,3'-Dibromo-2,2'-dimethoxy-1,1'-binaphthyl 98.0+%, TCI America™
SDP

Catalog No. D28011G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D28011G 1 g
D2801200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D28011G Supplier TCI America Supplier No. D28011G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 75714-59-9
Molecular Formula C22H16Br2O2
Molecular Weight (g/mol) 472.176
MDL Number MFCD04038415
InChI Key DFTUKDIMHCCQIT-UHFFFAOYSA-N
PubChem CID 394170
IUPAC Name 3-bromo-1-(3-bromo-2-methoxynaphthalen-1-yl)-2-methoxynaphthalene
SMILES COC1=C(C2=CC=CC=C2C=C1Br)C3=C(C(=CC4=CC=CC=C43)Br)OC

Specifications

Melting Point 185°C
Color White
Quantity 1 g
Formula Weight 472.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (R)-3,3′-Dibromo-2,2′-dimethoxy-1,1′-binaphthyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.