Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(-)-3,4,8,8a-Tetrahydro-8a-methyl-1,6(2H,7H)-naphthalenedione, 98%, Thermo Scientific™

Catalog No. AAL2005803
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2005803 1 g
AAL20058MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL2005803 Supplier Thermo Scientific Chemicals Supplier No. L2005803

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100348-93-4
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.23
MDL Number MFCD00063004
InChI Key DNHDRUMZDHWHKG-UHFFFAOYNA-N
Synonym r---8a-methyl-3,4,8,8a-tetrahydro-1,6 2h,7h-naphthalenedione, 8ar-8a-methyl-3,4,7,8-tetrahydro-2h-naphthalene-1,6-dione, 8ar-8a-methyl-3,4,8,8a-tetrahydronaphthalene-1,6 2h,7h-dione, 1,6 2h,7h-naphthalenedione, 3,4,8,8a-tetrahydro-8a-methyl-, 8ar, r-miescher-wieland ketone, r---9-methyl-5 10-octaline-1,6-dione, --r-6-methylbicyclo 4.4.0 dec-1-ene-3,7-dione, r--8a-methyl-3,4,8,8a-tetrahydro-1,6 2h,7h naphthalenendione, r---3,4,8,8a-tetrahydro-8a-methyl-1,6 2h,7h-naphthalenedione, r--8a-methyl-3,4,8,8a-tetrahydro-1,6 2h,7h-naphthalenedione
PubChem CID 736070
IUPAC Name (8aR)-8a-methyl-3,4,7,8-tetrahydro-2H-naphthalene-1,6-dione
SMILES CC12CCC(=O)C=C1CCCC2=O

Specifications

Melting Point 48°C to 50°C
Quantity 1 g
Beilstein 2050425
Formula Weight 178.23
Percent Purity 98%
Chemical Name or Material (R)-(-)-3,4,8,8a-Tetrahydro-8a-methyl-1,6(2H,7H)-naphthalenedione
TSCA No
Recommended Storage Keep cold
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.