Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(-)-(3,5-Dioxa-4-phosphacyclohepta[2,1-a;3,4-a']dinaphthalen-4-yl)dimethylamine 98.0+%, TCI America™
SDP

Catalog No. D49921G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D49921G 1 g
D49925G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D49921G Supplier TCI America Supplier No. D49921G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 157488-65-8
Molecular Formula C22H18NO2P
Molecular Weight (g/mol) 359.365
MDL Number MFCD03426988
InChI Key QCHAVHXSBZARBO-UHFFFAOYSA-N
PubChem CID 10893715
SMILES CN(C)P1OC2=C(C3=CC=CC=C3C=C2)C4=C(O1)C=CC5=CC=CC=C54

Specifications

Melting Point 227°C
Color White-Yellow
Quantity 1 g
Formula Weight 359.36
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material (R)-(-)-(3,5-Dioxa-4-phosphacyclohepta[2,1-a;3,4-a′]dinaphthalen-4-yl)dimethylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.