Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-3-Amino-3-phenyl-1-propanol 97.0+%, TCI America™
SDP

Catalog No. A29991G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A29991G 1 g
A2999200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A29991G Supplier TCI America Supplier No. A29991G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 170564-98-4
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.209
MDL Number MFCD01311791
InChI Key SEQXIQNPMQTBGN-SECBINFHSA-N
Synonym r-1-phenyl-3-propanolamine, r-3-amino-3-phenylpropan-1-ol, 3r-3-amino-3-phenylpropan-1-ol, r-beta-phenylalaninol, r-3-amino-3-phenyl-1-propanol, r-3-phenyl-beta-alaninol, r-3-amino-3-phenyl-propan-1-ol, r--phenylalaninol, pubchem13853
PubChem CID 7016858
IUPAC Name (3R)-3-amino-3-phenylpropan-1-ol
SMILES C1=CC=C(C=C1)C(CCO)N

Specifications

Melting Point 77°C
Color White-Yellow
Quantity 1 g
UN Number 3259
Formula Weight 151.21
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material (R)-3-Amino-3-phenyl-1-propanol
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.