Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-3-(Boc-amino)-3-(3-bromophenyl)propionic acid, 97%, Thermo Scientific™

Catalog No. AAH5203306
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5203306 5 g
AAH52033MD 250 mg
AAH5203303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH5203306 Supplier Thermo Scientific Chemicals Supplier No. H5203306

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 501015-16-3
Molecular Formula C14H18BrNO4
Molecular Weight (g/mol) 344.205
MDL Number MFCD03427962
InChI Key UTYRIGRZLZNTLR-LLVKDONJSA-N
Synonym r-n-boc-3-bromo-beta-phenylalanine, r-3-3-bromophenyl-3-tert-butoxycarbonyl amino propanoic acid, boc-r-3-amino-3-3-bromo-phenyl-propionic acid, r-3-boc-amino-3-3-bromophenyl propionic acid, boc-s-3-amino-3-3-bromophenyl-propionic acid, 3r-3-3-bromophenyl-3-tert-butoxycarbonyl amino propanoic acid, boc-r-3-amino-3-3-bromophenyl-propionic acid, boc-d-?-phe 3-br-oh, boc-d-beta-phe 3-br-oh
PubChem CID 2761727
IUPAC Name (3R)-3-(3-bromophenyl)-3-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CC(=O)O)C1=CC(=CC=C1)Br

Specifications

Melting Point 77°C to 79°C
Quantity 5 g
Optical Rotation +44° (c=1 in Ethanol)
Formula Weight 344.2
Percent Purity 97%
Chemical Name or Material (R)-3-(Boc-amino)-3-(3-bromophenyl)propionic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.