Learn More
(R)-3-(Boc-amino)-3-(3-methoxyphenyl)propionic acid, 97%, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals, petrochemicals, and dyestuff.
Solubility
Slightly soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Specifications
| Melting Point | 106°C to 108°C |
| Quantity | 5 g |
| Optical Rotation | +40° (c=1 in Ethanol) |
| Formula Weight | 295.34 |
| Percent Purity | 97% |
| Chemical Name or Material | (R)-3-(Boc-amino)-3-(3-methoxyphenyl)propionic acid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.