Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(+)-3-Butyn-2-ol, 99%

Catalog No. AAL1888003
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1888003 1 g
AAL18880MD 250 mg
AAL1888006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1888003 Supplier Thermo Scientific Chemicals Supplier No. L1888003
Add to Cart

CAS: 42969-65-3 | C4H6O | 70.091 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42969-65-3
Molecular Formula C4H6O
Molecular Weight (g/mol) 70.091
MDL Number MFCD00211237
InChI Key GKPOMITUDGXOSB-SCSAIBSYSA-N
Synonym r-+-3-butyn-2-ol, r-3-butyn-2-ol, 2r-but-3-yn-2-ol, r-but-3-yn-2-ol, 2r-3-butyn-2-ol, 3-butyn-2-ol, 2r, r-+-1-butyn-3-ol, +-3-butyn-2-ol, pubchem20211, r---3-butyne-2-ol
PubChem CID 638102
IUPAC Name (2R)-but-3-yn-2-ol
SMILES CC(C#C)O

Specifications

Density 0.891
Boiling Point 108°C to 109°C
Flash Point 28°C (82°F)
Refractive Index 1.426
Quantity 1 g
UN Number UN2929
Beilstein 3587272
Optical Rotation +47° (Neat)
Formula Weight 70.09
Percent Purity 99%
Chemical Name or Material (R)-(+)-3-Butyn-2-ol
Show More Show Less
Hazard Category H226-H300+H310+H330-H315-H319-H335
Hazard Statement GHS H Statement
H300-H310-H226-H315-H319-H335
Fatal if swallowed.
Fatal in contact with skin.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P262-P264b-P270-P271-P280-P284-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, FLAMMABLE, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.