Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(-)-4-Benzyl-3-propionyl-2-oxazolidinone 98.0+%, TCI America™
SDP

Catalog No. B216525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B216525G 25 g
B21651G 1 g
B21655G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B216525G Supplier TCI America Supplier No. B216525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 131685-53-5
Molecular Formula C13H15NO3
Molecular Weight (g/mol) 233.267
MDL Number MFCD00269687
InChI Key WHOBYFHKONUTMW-LLVKDONJSA-N
PubChem CID 10966403
IUPAC Name (4R)-4-benzyl-3-propanoyl-1,3-oxazolidin-2-one
SMILES CCC(=O)N1C(COC1=O)CC2=CC=CC=C2

Specifications

Melting Point 45°C
Color White-Yellow
Quantity 25 g
Formula Weight 233.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (R)-(-)-4-Benzyl-3-propionyl-2-oxazolidinone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.