Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-4-Carboxyphenylglycine, Tocris Bioscience™

Catalog No. 032210R
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
03-221-0R 10 mg
03-225-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 03-221-0R Supplier R&D Systems Supplier No. 0322/10

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Moderately potent NMDA antagonist

Moderately potent NMDA receptor antagonist.

Racemate and S-enantiomer also available.

Chemical Identifiers

CAS 134052-68-9
Molecular Formula C9H9NO4
Molecular Weight (g/mol) 195.174
InChI Key VTMJKPGFERYGJF-SSDOTTSWSA-N
Synonym r-4-carboxyphenylglycine, r-4-amino carboxy methyl benzoic acid, tocris-0321, tocris-0322, r-4cpg, r-alpha-amino-alpha-4-carboxyphenyl acetic acid, 4-1r-1-amino-2-hydroxy-2-oxoethyl benzoic acid
PubChem CID 6604711
IUPAC Name 4-[(R)-amino(carboxy)methyl]benzoic acid
SMILES C1=CC(=CC=C1C(C(=O)O)N)C(=O)O

Specifications

Quantity 10 mg
Formula Weight 195.17
Percent Purity >98%
Chemical Name or Material (R)-4-Carboxyphenylglycine
Recommended Storage Store at room temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.