Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(+)-5,5',6,6',7,7',8,8'-Octahydro-1,1'-bi-2-naphthol 99.0+%, TCI America™
SDP

Catalog No. O028225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O028225G 25 g
O02821G 1 g
O02825G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. O028225G Supplier TCI America Supplier No. O028225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 65355-14-8
Molecular Formula C20H22O2
Molecular Weight (g/mol) 294.394
MDL Number MFCD02093485
InChI Key UTXIFKBYNJRJPH-UHFFFAOYSA-N
Synonym s---5,5',6,6',7,7',8,8'-octahydro-1,1'-bi-2-naphthol, r-+-5,5',6,6',7,7',8,8'-octahydro-1,1'-bi-2-naphthol, r-5,5',6,6',7,7',8,8'-octahydro 1,1'-binaphthalene-2,2'-diol, s-5,5',6,6',7,7',8,8'-octahydro-1,1'-binaphthalene-2,2'-diol, 5,5',6,6',7,7',8,8'-octahydro-1,1'-binaphthalene-2,2'-diol, 5,5',6,6',7,7',8,8'-octahydro-1,1'-bi-2-naphthol, 5,5',6,6',7,7',8,8'-octahydro-bi-2-naphthol, r-5,5',6,6',7,7',8,8'-octahydro-1,1'-bi-2-naphthol, s---2,2'-dihydroxy-5,5',6,6',7,7',8,8'-octahydro-1,1'-binaphthyl, 1-2-hydroxy-5,6,7,8-tetrahydronaphthalen-1-yl-5,6,7,8-tetrahydronaphthalen-2-ol
PubChem CID 3694111
IUPAC Name 1-(2-hydroxy-5,6,7,8-tetrahydronaphthalen-1-yl)-5,6,7,8-tetrahydronaphthalen-2-ol
SMILES C1CCC2=C(C1)C=CC(=C2C3=C(C=CC4=C3CCCC4)O)O

Specifications

Melting Point 167°C
Color White
Quantity 25 g
Formula Weight 294.39
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (R)-(+)-5,5′,6,6′,7,7′,8,8′-Octahydro-1,1′-bi-2-naphthol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.