Learn More
Strem, An Ascensus Company (R)-(+)-5 5 -BIS DI(3 5--0.25G
SDP
Supplier:  Strem, An Ascensus Company 150478250MG
Encompass
(R)-(+)-5,5'-Bis[di(3,5-xylyl)phosphino]-4,4'-bi-1,3-benzodioxole, min. 98% (R)-DM-SEGPHOS; Size: 250mg; [850253-53-1]; C46H44O4P2; Mol Wt:722.79; Color & Form: off white to pale yellow pwdr.
Catalog No. 50-901-09914
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.