Learn More
Abcam (R)-(+)-8-Hydroxy-DPAT Hydrobromide, 5-HT1A Receptor Agonist, Solid, 10 mg
Description
- Selective full 5-HT1A receptor agonist
- More active enantiomer of 8-OH-DPAT
- Displays greater efficacy at 5-HT1A
- Displays moderate affinity at 5-HT7
- Inhibits amphetamine induced dopamine release
- Molecular weight: 328.29 Da
- Purity: >98%
- Form: Solid
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Content And Storage | Store at 4°C. |
| Shipping Condition | Ambient |
| Target Molecule | 5-HT1A Receptor, 5-HT7 Receptor |
| Buffer | 100% (R)-(+)-8-Hydroxy-DPAT hydrobromide |
| Form | Solid |
| Formulation | 100% (R)-(+)-8-Hydroxy-DPAT hydrobromide |
| Quantity | 10 mg |
| For Use With (Application) | 5-HT1A receptor agonist, 5-HT7 receptor ligand, dopamine release inhibition |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.