Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(-)-alpha-Methoxyphenylacetic acid, 99%

Catalog No. AAL0871903
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0871903 1 g
AAL08719MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0871903 Supplier Thermo Scientific Chemicals Supplier No. L0871903
Add to Cart

CAS: 3966-32-3 | C9H10O3 | 166.176 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3966-32-3
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00004250
InChI Key DIWVBIXQCNRCFE-MRVPVSSYSA-N
Synonym r---alpha-methoxyphenylacetic acid, r-2-methoxy-2-phenylacetic acid, 2r-2-methoxy-2-phenylacetic acid, r-methoxyphenylacetic acid, r-methoxy phenyl acetic acid, methoxy phenyl acetic acid #, r---.alpha.-methoxyphenylacetic acid, benzeneacetic acid, .alpha.-methoxy-, r, o-methyl-d-mandelic acid, ksc496q9j
PubChem CID 2724294
IUPAC Name (2R)-2-methoxy-2-phenylacetic acid
SMILES COC(C1=CC=CC=C1)C(=O)O

Specifications

Melting Point 63°C to 68°C
Boiling Point 164°C to 166°C (18 mmHg)
Quantity 1 g
Beilstein 3589257
Optical Rotation −146° (c=0.5 in Ethanol)
Formula Weight 166.18
Percent Purity 99%
Chemical Name or Material (R)-(-)-alpha-Methoxyphenylacetic acid
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 223-580-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.