Learn More
Medchemexpress LLC (R)-alpha-tocotrienol | 58864-81-6 | MFCD11045308 | 98.8% | 424.66 g/mol | C29H44O2 | 5 MG

Supplier: Medchemexpress LLC HY1294595MG
α-Tocotrienol (CAS 58864-81-6) is an isoform of vitamin E provided as a high-purity analytical standard for biochemical and analytical applications. It is supplied as a colorless to light yellow oil, characterized by molecular formula C29H44O2 and molecular weight 424.66 g/mol, and tested for identity and purity by HPLC and NMR.
- High purity suitable for analysis and research (HPLC 98.81%).
- Characterized by 1H NMR consistent with structure.
- Colorless to light yellow oil physical appearance.
- Molecular formula C29H44O2 and molecular weight 424.66 g/mol.
- Stable when stored sealed at -20°C; in solvent: -80°C (6 months) or -20°C (1 month).
- Useful as an analytical standard and research reagent for antioxidant and vitamin E studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.