Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam (R)-Baclofen, GABAB Agonist, Solid, 10 mg

Catalog No. 502707676
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-7676 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-7676 Supplier Abcam Supplier No. AB12032510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

More active enantiomer of the selective GABAB agonist (R,S)-Baclofen, with >98% purity.

  • Molecular Weight: 213.66 Da
  • Purity: >98%
  • Description: More active enantiomer of selective GABAB agonist (R,S)-Baclofen (ab120149)
  • Solubility: Soluble in 1 mL water
  • Form: Solid
  • CAS Number: 69308-37-8
  • Storage: Store at ambient temperature

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Shipping Condition Ambient
Target Molecule GABAB Receptor
Content And Storage Store at ambient temperature.
Form Solid
Formulation 100% (R)-Baclofen
Quantity 10 mg
For Use With (Application) GABAB receptor activation
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.