Learn More
Abcam (R)-Baclofen, GABAB Agonist, Solid, >98% Purity, 50 mg
Description
- Compound type: Selective GABAB agonist
- Activity: More active enantiomer of (R,S)-Baclofen
- Purity: >98%
- Molecular weight: 213.66 Da
- Form: Solid
- CAS number: 69308-37-8
- Storage: Store at ambient temperature
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Shipping Condition | Ambient |
| Buffer | 100% (R)-Baclofen |
| Form | Solid |
| Target Molecule | GABAB Receptor |
| Content And Storage | Store at ambient temperature. |
| Quantity | 50 mg |
| For Use With (Application) | GABAB receptor agonist |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.