Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(+)-beta-Methylphenethylamine 98.0+%, TCI America™
SDP

Catalog No. M174625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M174625G 25 g
M17465G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M174625G Supplier TCI America Supplier No. M174625G

Chemical Identifiers

CAS 28163-64-6
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00216740
InChI Key AXORVIZLPOGIRG-QMMMGPOBSA-N
Synonym (R)-(+)-2-Phenyl-1-propylamine
PubChem CID 1547949
IUPAC Name (2R)-2-phenylpropan-1-amine
SMILES CC(CN)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 102°C
Quantity 25 g
UN Number 2735
Formula Weight 135.21
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material (R)-(+)-beta-Methylphenethylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.