Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(+)-BINAP, TCI America™
SDP

Catalog No. B140625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1406-25G 25 g
B1406-5G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B1406-25G Supplier TCI America Supplier No. B140625G
Add to Cart
Add to Cart

Specifications

CAS 76189-55-4
Melting Point 242°C
Color White-Yellow
Molecular Formula C44H32P2
MDL Number MFCD00010805
Quantity 25 g
Synonym binap, r-+-2,2 '-bis diphenylphosphino-1,1 '-binaphthyl, 2,2 '-bis diphenylphosphino-1,1 '-binaphthyl, s-binap, s---binap
InChI Key MUALRAIOVNYAIW-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=CC=C7)C8=CC=CC=C8
IUPAC Name [1-(2-diphenylphosphanylnaphthalen-1-yl)naphthalen-2-yl]-diphenylphosphane
Molecular Weight (g/mol) 622.688
PubChem CID 634876
Formula Weight 622.69
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.