Learn More
Abcam (R)-Butaprost, EP2 Agonist, >98% Purity, Liquid, 1 mg
Description
- Selective EP2 agonist
- PGE2 structural analog
- Inhibits chemotaxis (EC50 = 106 nM, maximal effect of PGE2)
- Increases neutrophil cyclic AMP levels
- Molecular Weight: 408.6 Da
- Purity: >98%
- Form: Liquid
- Storage buffer: Constituents: 99% Methyl acetate, 1% (R)-Butaprost
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Buffer | 99% methyl acetate, 1% (R)-Butaprost |
| Content And Storage | Store at -20°C. |
| Form | Liquid |
| Shipping Condition | Ambient |
| Substrate Properties | Chemical Substrate |
| Target Molecule | EP2 |
| Formulation | 99% Methyl acetate, 1% (R)-Butaprost |
| Quantity | 1 mg |
| For Use With (Application) | Inhibits chemotaxis, increases neutrophil cyclic AMP levels |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.