Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam (R)-Butaprost, EP2 Agonist, >98% Purity, Liquid, 5 mg

Catalog No. 502705681
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-5681 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-5681 Supplier Abcam Supplier No. AB1444765MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Selective EP2 agonist and PGE2 structural analog that inhibits chemotaxis.

  • Selective EP2 agonist
  • PGE2 structural analog
  • Inhibits chemotaxis (EC50 = 106 nM, maximal effect of PGE2)
  • Increases neutrophil cyclic AMP levels
  • Purity >98%
  • Molecular weight: 408.6 Da

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Prostaglandin E2 Receptor (EP2)
Content And Storage Store at -20°C.
Buffer 99% methyl acetate, 1% (R)-Butaprost
Form Liquid
Shipping Condition Ambient
Formulation 99% Methyl acetate, 1% (R)-Butaprost
Quantity 5 mg
For Use With (Application) Chemotaxis inhibition, Neutrophil cyclic AMP elevation
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.