Learn More
Description
Includes
1.25mL 4X Agarose cleaning buffer; 1mL 5X Agarose wash buffer; 1mL 5X Agarose storage buffer
With proper cleaning and storage, agarose resins can be reused for at least 5 to 10 applications
Specifications
Specifications
| Form | Liquid |
| Quantity | 1 Kit: 1.25 mL 4X Agarose cleaning buffer, 1mL 5X Agarose wash buffer, 1 mL 5X Agarose storage buffer |
| Storage Requirements | Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Product Type | Agarose Cleaning and Storage Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
