Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Avidin-Fluorescein
SDP

Catalog No. F0030 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0030 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. F0030 Supplier R&D Systems™ Supplier No. F0030

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Applications: Flow Cytometry

  • Optimized for use with monoclonal antibody-biotin reagents
  • Av-FITC is provided in a ready-to-use liquid format containing 0.15M NaCl, 25mM HEPES, 0.5% BSA, and 0.1% NaN3
  • FITC molecule can be excited by laser light at 488nm and has an optimal emission at wavelength at 525nm

Contains sodium azide as a preservative: sodium azide may react with lead and copper plumbing to form explosive metal azides. Flush with large volumes of water during disposal.

Applications:

Flow Cytometry

Specifications

Quantity 100 Tests
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Line R&D Systems
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.