Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ AZ 191
GREENER_CHOICESDP

Catalog No. 523250
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
523250 50 mg
523210 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 523250 Supplier Tocris Bioscience™ Supplier No. 5232/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Potent DYRK1B inhibitor

AZ 191 is a potent DYRK1B inhibitor (IC50 values are 17, 88 and 1890 nM for DYRK1B, DYRK1A and DYRK2 respectively). Inhibits DYRK1B-induced Gi phase cell cycle arrest in Panc-1 cells in vitro.

Specifications

Chemical Name or Material AZ 191
CAS 1594092-37-1
Synonym N-[2-Methoxy-4-(4-methyl-1-piperazinyl)phenyl]-4-(1-methyl-1H-pyrrolo[2,3-c]pyridin-3-yl)-2-pyrimidinamine
Quantity 50 mg
Solubility Soluble to 100mM in DMSO and to 20mM in ethanol
Molecular Formula C24H27N7O
Formula Weight 429.52
Physical Form Solid
Product Type Bioactive Small Molecule
Purity >98%

Research Use Only. Not For Human or Veterinary Use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.