Learn More
R&D Systems™ Boc-QAR-AMC Fluorogenic Peptide Substrate
SDP
Catalog No. ES014 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
Catalog No. Quantity
ES014 10 mg
1 options
This item is not returnable. View return policy
Catalog No. ES014 Supplier R&D Systems™ Supplier No. ES014
Customers require a valid business account associated to their profile to access pricing and ordering options.
This item is not returnable. View return policy
Substrate for matriptase, spinesin, and prostasin
  • Hydrolysis of Arg-AMC amide bond releases AMC, a highly fluorescent group
  • A fluorescence plate reader with excitation at 380nm and emission at 460nm is recommended for the measurement of the enzymatic activity
  • Purity >98%
  • Excellent substrate for several serine proeases
Specifications
Quantity 10 mg
Product Type Boc-Gln-Ala-Arg-AMC HCl
Storage Requirements Samples are stable for up to twelve months from date of receipt at -20° C to -70° C. The substrate can be aliquoted and stored -20° C to -70° C in a manual defrost freezer for three months. Avoid repeated freeze-thaw cycles. Protect from exposure to direct light.