Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ C 87

Catalog No. 548450
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
548450 50 mg
548410 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 548450 Supplier Tocris Bioscience™ Supplier No. 5484/50
This item is not returnable. View return policy

TNF-α inhibitor

C 87 is a TNF-α inhibitor (Kd = 110 nM for hTNF-α). Inhibits TNF-α-induced cytotoxicity in L929 fibroblast cells. Attenuates inflammation and increases survival in a mouse hepatitis model.

Specifications

Chemical Name or Material C 87
CAS 332420-90-3
Synonym 3-Phenyl-1-(4-phenyl-2-thiazolyl)-1H-pyrazole-4,5-dione 4-[2-(4-chloro-3-nitrophenyl)hydrazone]
Quantity 50 mg
Solubility Soluble to 20mM in DMSO
Molecular Formula C24H15ClN6O3S
Formula Weight 502.93
Physical Form Solid
Product Type Bioactive Small Molecule
Purity >98%

Research Use Only. Not For Human or Veterinary Use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.