Learn More
Description
- This kit is designed for the detection and enumeration of canine IL-2 secreting cells.
- Assay Type: ELISpot (quantitative sandwich ELISA technique)
- Format: 96-well PVDF-backed microplate
- Sample Type: Whole cells
- Features: High sensitivity
- Required Equipment: Dissection microscope or an ELISpot reader
- Components: Canine IL-2 microplate, biotinylated detection antibody, streptavidin conjugated to alkaline phosphatase
Specifications
Specifications
| Cross Reactivity | Dog |
| Includes | Six 96-well plates coated w/Ab for canine IL-2, Biotinylated detection Ab, Streptavidin-AP, Buffers, BCIP/NBT, Positive control |
| Quantity | 6 x 96-well Plates |
| Sample Type | Whole Cells |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Form | Liquid |
| Product Line | R&D Systems |
| For Use With (Application) | Diagnostics |
| Product Type | Canine IL-2 ELISpot Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
