Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Cultrex Basement Membrane Extract, Pathclear (w/ Phenol Red)
SDP

Catalog No. 343201001P Shop All Novus Products
Change view
Click to view available options
Quantity:
5 mL
2 x 5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
343201001P 2 x 5 mL
343200501P 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 343201001P Supplier R&D Systems™ Supplier No. 343201001P
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

soluble form of basement membrane purified from Engelbreth-Holm-Swarm (EHS) tumor

Functional Assay: Tube formation assay - BME promotes formation of capillary-like structures by human (HBMVEC; HUVEC) or mouse (SVEC4-10) endothelial cells. Gelling Assay: BME gels in less than 30 minutes at 37degreesC, and maintains the gelled form in culture medium for a minimum of 14 days at 37degreesC.

Specifications

Concentration 8-12 mg/mL (by Lowry assay)
For Use With (Application) Bioactivity
Product Line R&D Systems
Quantity 2 x 5 mL
Endotoxin Level ≤8EU/mL
Source Murine Engelbreth-Holm-Swarm (EHS) tumor.
Content And Storage Product is stable for a minimum of 3 months from date of shipment when stored at -20° C in a manual defrost freezer. For optimal stability, store at -80° C. Avoid freeze-thaw cycles.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.