Learn More
R&D Systems™ Cultrex RGF Basement Membrane Extract, Type 2
Description
Specifications
Specifications
| Concentration | 8-12 mg/mL (by Lowry assay) |
| For Use With (Application) | Bioactivity |
| Quantity | 5 mL |
| Source | Murine Engelbreth-Holm-Swarm (EHS) tumor. |
| Content And Storage | Product is stable for a minimum of 3 months from date of shipment when stored at -20°C in a manual defrost freezer. For optimal stability, store at -80°C. Avoid freeze-thaw cycles. Storage Buffer: Dulbecco’s Modified Eagle’s medium without phenol red, with 10 μg/mL gentamicin sulfate. Sterility Testing: PathClear™ - Negative by PCR test for mycoplasma; 17 bacterial and virus strains typically included in mouse antibody production (MAP) testing, plus 13 additional murine infectious agents including LDEV, for a total of 31 organisms and viruses. No bacterial or fungal growth detected after incubation (at 37°C for 14 days following USP sterility testing guidelines.) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.