Learn More
R&D Systems™ Cultrex Basement Membrane Extract, Type 3 (w/ Phenol Red)
Description
Specifications
Specifications
| Concentration | 8-12 mg/mL (by Lowry assay) |
| For Use With (Application) | Bioactivity |
| Quantity | 5 mL |
| Source | Murine Engelbreth-Holm-Swarm (EHS) tumor. |
| Content And Storage | Product is stable for a minimum of 3 months from date of shipment when stored at -20° C in a manual defrost freezer. For optimal stability, store at -80° C. Avoid freeze-thaw cycles. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.