Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ESI 09
GREENER_CHOICE

Catalog No. p-7082167
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
477310 10 mg
477350 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 477310 Supplier Tocris Bioscience™ Supplier No. 4773/10
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Epac inhibitor

ESI 09 is an EPAC inhibitor. Inhibits EPAC-mediated insulin release from pancreatic β cells. Selective for EPAC over PKA. ESI 09 decreases migration and invasion of AsPc-1 and PANC-1 pancreatic cancer cells in vitro.

Specifications

Chemical Name or Material EPAC Inhibitors
CAS 263707-16-0
Synonym α-[(2-(3-Chlorophenyl)hydrazinylidene]-5-(1,1-dimethylethyl)-β-oxo-3-isoxazolepropanenitrile
Quantity 10 mg
Solubility Soluble to 100mM in DMSO and to 20mM in ethanol
Molecular Formula C16H15ClN4O2
Formula Weight 330.77
Physical Form Solid
Product Type Bioactive Small Molecule
Purity >98%

Research Use Only. Not For Human or Veterinary Use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.