Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Flow Cytometry Mouse Lyse Buffer (10X), R&D Systems™
SDP

Catalog No. FC003 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FC003 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FC003 Supplier R&D Systems Supplier No. FC003

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Applications: Flow Cytometry

  • Gently lyses mouse erythrocytes

Applications:

Flow Cytometry

Specifications

Product Type Lysis Buffer
Content And Storage Store the unopened product at 2 - 8° C. Do not use past expiration date.
Formulation Spleen or whole blood samples
Quantity 50mL
Stability Upto 6 Months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.