Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Flow Cytometry Permeabilization/Wash Buffer I (1X), R&D Systems™
GSA_VASDP

Catalog No. FC005 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FC005 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FC005 Supplier R&D Systems Supplier No. FC005

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Applications: Flow Cytometry

  • Designed for intracellular flow cytometry applications with nonconjugated or fluorochrome-conjugated antibodies

Applications:

Flow Cytometry

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Product Type Permeabilization/Wash Buffer
Formulation Sodium azide 0.05%
Content And Storage Store the unopened product at 2 - 8° C. Do not use past expiration date.
Quantity 250mL
Stability Upto 6 Months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.