Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Flow Cytometry Staining Buffer (1X), R&D Systems™
SDP

Catalog No. FC001 Shop All Novus Products
Change view
Click to view available options
Quantity:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FC001 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FC001 Supplier R&D Systems Supplier No. FC001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Applications: Flow Cytometry

  • Optimized for immunofluorescent staining of single-cell suspensions and subsequent dilution, suspension, washing and storage of cells intended for flow cytometry
  • Supplemented with Bovine Serum Albumin (BSA) and the metabolic inhibitor sodium azide

Applications:

Flow Cytometry

Specifications

Content And Storage Store the unopened product at 2 - 8° C. Do not use past expiration date.
Formulation Sodium azide 0.1% and bovine serum albumin
Quantity 500mL
Stability Upto 6 Months
Product Type Staining Buffer
Product Line R&D Systems
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.