Learn More
R&D Systems™ Histidine-tagged Protein Purification Resin
Catalog No. IP999
Change view
Click to view available options
2mL
Catalog No.
IP999 2mL
1 options
This item is not returnable. View return policy
Catalog No. IP999 Supplier R&D Systems™ Supplier No. IP999
Add to Cart
Add to Cart
This item is not returnable. View return policy
Designed for the immunopurification of His-tagged proteins in a single step
Consecutive histidine residues (usually 6 to 10 in. length) are often inserted into the amino acid sequences of recombinant proteins. The resulting His-tagged proteins can be detected or purified by using anti-polyHis antibodies.
  • Agarose beads covalently linked to a murine monoclonal antibody specific for polyhistidine-tagged peptides
  • The resin can also be used in small quantities to immunoprecipitate His-tagged proteins
  • Each mL of packed resin can bind more than 0.5mg of His-tagged proteins (protocol-dependent)
Specifications
Content And Storage Store the unopened product at 2 - 8° C. Do not use past expiration date.
Quantity 2 mL
Type Resin