Learn More
R&D Systems™ Human 19S Proteasome FLAG-UCHL5 (C88A) Recombinant Protein
Description
Specifications
Specifications
| Formulation | X mg/mL (X μM) in 20 mM HEPES pH 7.5, 20 mM NaCl, 20% Glycerol (v/v), 1 mM DTT |
| Molecular Weight (g/mol) | 700 kDa |
| Quantity | 25 μg |
| Source | Human embryonic kidney cell,HEK293-derived human 19S Proteasome protein |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening. |
| Product Type | 19S Proteasome FLAG-UCHL5 (C88A) |
| Conjugate | Unconjugated |
| Species | Human |
| Recombinant | Recombinant |
| Purity or Quality Grade | 70%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.