Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human 20S Proteasome Protein
SDP

Catalog No. E360050 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E360050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. E360050 Supplier R&D Systems™ Supplier No. E360050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

The Human 20S Proteasome is able to degrade substrates in an ATP-independent manner. Reaction conditions will need to be optimized for each specific application. We recommend an initial Human 20S Proteasome concentration of 0.5-5 nM.

Specifications

Formulation Supplied as a 0.2 μm filtered solution in HEPES, NaCl and DTT.
Molecular Weight (g/mol) 700 kDa
Quantity 50 μg
Source Human erythrocyte-derived 20 S Proteasome protein The Human 20 S Proteasome protein has been purified from human erythrocytes, which have been screened and are negative for hepatitis B surface antigen, antibodies to hepatitis C virus, HIV type 1 antigens, and antibodies to HIV type 1 and 2.
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.
Product Type 20S Proteasome
Conjugate Unconjugated
Species Human
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.