Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Apo-Transferrin Protein
SDP

Catalog No. 3188AT100MG Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
3188AT100MG 100 mg
3188AT001G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 3188AT100MG Supplier R&D Systems™ Supplier No. 3188AT100MG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured in a serum-free cell proliferation assay using CHO-K1 cells. The ED50 for this effect is <6.00 μg/mL.

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in deionized water.
Name Apo-Transferrin
Quantity 100 mg
Source Human plasma-derived Apo-Transferrin protein The human plasma used for the isolation of this product were certified by the supplier to be negative for HIV I and II antibodies, Hepatitis C antibody and Hepatitis B antigen at the time of shipment. Human blood products should always be treated in accordance with universal handling precautions.
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Conjugate Unconjugated
Purity or Quality Grade 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.