Learn More
R&D Systems™ Recombinant Human APRIL/TNFSF13 (HEK293-expressed) Protein
Shop All Novus Biologicals Products

Description
Specifications
Specifications
| Accession Number | O75888 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in Tris-HCl and NaCl with BSA as a carrier protein. |
| Gene ID (Entrez) | 8741 |
| Molecular Weight (g/mol) | M.W. (Observed): 26 kDa, reducing conditions; M.W. (theoretical): 21.7 kDa |
| Name | APRIL/TNFSF13 |
| Quantity | 10 μg |
| Source | Human embryonic kidney cell, HEK293-derived human APRIL/TNFSF13 protein HA (YPYDVPDYA) GCN4-IZ GGGSGGGSGGGS Human April (Ala105-Leu250) (N-terminus) Accession # O75888 (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Structural Form | Noncovalently-linked homotrimer |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.