Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human APRIL/TNFSF13 (HEK293-expressed) Protein

Catalog No. 5860AP010CF
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2 μm filtered solution in Tris-HCl and NaCl with BSA as a carrier protein.
Lyophilized from a 0.2 μm filtered solution in Tris-HCl and NaCl.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation
5860AP010CF Lyophilized from a 0.2 μm filtered solution in Tris-HCl and NaCl.
5860AP010 Lyophilized from a 0.2 μm filtered solution in Tris-HCl and NaCl with BSA as a carrier protein.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 5860AP010CF Supplier R&D Systems™ Supplier No. 5860AP010/CF
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured in a cell proliferation assay using anti-IgM stimulated mouse B cells. The ED50 for this effect is 5-25 ng/mL in the presence of goat anti-mouse IgM.

Specifications

Accession Number O75888
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in Tris-HCl and NaCl.
Gene ID (Entrez) 8741
Molecular Weight (g/mol) M.W. (Observed): 26 kDa, reducing conditions; M.W. (theoretical): 21.7 kDa
Name APRIL/TNFSF13
Quantity 10 μg
Source Human embryonic kidney cell, HEK293-derived human APRIL/TNFSF13 protein HA (YPYDVPDYA) GCN4-IZ GGGSGGGSGGGS Human April (Ala105-Leu250) (N-terminus) Accession # O75888 (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Structural Form Noncovalently-linked homotrimer
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol TNFSF13
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in PBS.
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Protein APRIL/TNFSF13
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.