Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human CCL15/MIP-1 delta 92 aa Recombinant Protein

Catalog No. 363MG025
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation
363MG025 Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
363MG025CF Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 363MG025 Supplier R&D Systems™ Supplier No. 363MG025
This item is not returnable. View return policy

Source: E. coli-derived, Gln22-Ile113
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 10 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 6359
Regulatory Status: RUO

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
Gene ID (Entrez) 6359
Molecular Weight (g/mol) 10 kDa
Name CCL15/MIP-1 delta 92 aa
Quantity 25 μg
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.